missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
Specifications
Specifications
Density | 1.04 |
Boiling Point | 79°C (11 mmHg) |
Flash Point | 76°C (169°F) |
Refractive Index | 1.532 |
Quantity | 1 g |
UN Number | UN2810 |
Synonym | benzenamine, 4-fluoro-n-methyl, unii-88f75yis0f, n-methyl-4-fluoroaniline, acmc-1aem5, 4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, benzenamine,4-fluoro-n-methyl |
Formula Weight | 125.15 |
Percent Purity | 97% |
Chemical Name or Material | 4-Fluoro-N-methylaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'