missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals

Product Code. 11347376
Click to view available options
Quantity:
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5441-51-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00001644
InChI Key OKSDJGWHKXFVME-UHFFFAOYSA-N
Synonym 4-ethylcyclohexanone, cyclohexanone, 4-ethyl, 4-ethyl cyclohexanone, 4-ethyl-cyclohexanone, 1-ethyl-4-oxocyclohexane, trans-4-ethylcyclohexanone, acmc-1aru9, ksc273s8l, 5-ethyl-2-oxocyclohexyl
PubChem CID 79506
IUPAC Name 4-ethylcyclohexan-1-one
SMILES CCC1CCC(=O)CC1

Specifications

Density 0.895
Boiling Point 192°C to 194°C
Flash Point 63°C (145°F)
Odor Characteristic
Refractive Index 1.4515
Quantity 100 g
Beilstein 1924964
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 4-Ethylcyclohexanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'