missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 4-Epitetracycline hydrochloride, can be used as a secondary standard
149.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- 4-Epitetracycline hydrochloride is a metabolite derived from the degradation of tetracycline compounds and has been shown to display antibiotic activity
- This compound blocks protein synthesis by binding to the 30S ribosomal subunit and interfering with the binding of aminoacyl-tRNA to the mRNA-ribosome complex
Applications
- This semi-synthesized compound derived from actinomycetes can decrease the activity of the Tet repressor effector in animal models
- In vivo experiments revealed that 4-epitetracycline hydrochloride can induce a toxic response
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 217.0°C |
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 80% min. (as is,calculated) |
Packaging | Glass bottle |
Quantity | 100 mg |
Specific Rotation | − 322.00 |
Solubility Information | Solubility in water: 100g/L. Other solubilities: 100g/L methanol |
Formula Weight | 480.89 |
Percent Purity | ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC) |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- 4-Epitetracycline hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- H361d-Suspected of damaging the unborn child.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'