missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(5-isoxazolyl)phenol, 97%, Thermo Scientific™

Product Code. 11382429
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86176-56-9
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD00218660
InChI Key QCDHHUJPNPOGKY-UHFFFAOYSA-N
Synonym 4-chloro-2-isoxazol-5-yl-phenol, 4-chloro-6-2h-1,2-oxazol-5-ylidene cyclohexa-2,4-dien-1-one, 4-chloro-2-1,2-oxazol-5-yl phenol, aajxjlyuwkmqno-uhfffaoysa-n, 2-isoxazole-5-yl-4-chlorophenol, 5-2-hydroxy-5-chlorophenyl isoxazole
PubChem CID 6819866
IUPAC Name 4-chloro-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNO2)C=C1Cl

Specifications

Melting Point 200°C to 204°C
Quantity 25 g
Formula Weight 195.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(5-isoxazolyl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'