Learn More
Applied Biosystems™ 3130/3100-Avant Genetic Analyzer 4-Capillary Array, 80 cm
Use for fragment analysis and sequencing. Usage per array is 100 runs.
Brand:  Applied Biosystems™ 4333465
Product Code. 16335292
Description
Description
This is a 4-capillary array (internally uncoated) for the 3130 and 3100 Avant™ Genetic Analyzer.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 array
80 cm
Room Temperature
3100-Avant Genetic Analyzer, 3130 Genetic Analyzer
Store the array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide.
We guarantee the performance of the 3130 Capillary arrays up to 100 runs and within 1 year of purchase.
Avant™
170 min.
Sequencing, Fragment Analysis
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ 3130/3100-Avant Genetic Analyzer 4-Capillary Array, 80 cm > 80cm Capillary Length