missing translation for 'onlineSavingsMsg'
Learn More

4-Bromotriphenylamine, 99%, Thermo Scientific Chemicals

Product Code. 15426798
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36809-26-4
Molecular Formula C18H14BrN
Molecular Weight (g/mol) 324.221
MDL Number MFCD01851266
InChI Key SQTLUXJWUCHKMT-UHFFFAOYSA-N
Synonym 4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
PubChem CID 642838
IUPAC Name 4-bromo-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 108°C to 112°C
Quantity 25 g
Formula Weight 324.21
Percent Purity 99%
Chemical Name or Material 4-Bromotriphenylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'