missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromoacetyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 15444997
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5349-17-7
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
MDL Number MFCD02681893
InChI Key RGALBQILADNMKA-UHFFFAOYSA-N
Synonym 4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr
PubChem CID 2776239
IUPAC Name 2-bromo-1-pyridin-4-ylethanone;hydrobromide
SMILES C1=CN=CC=C1C(=O)CBr.Br

Specifications

Melting Point ∽200°C (decomposition)
Odor Weak
Quantity 5 g
UN Number UN3261
Solubility Information Soluble in water.
Formula Weight 280.95
Percent Purity 98%
Chemical Name or Material 4-(Bromoacetyl)pyridine hydrobromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'