Learn More
4-Bromo-N,N-dimethylaniline, 98+%, Thermo Scientific Chemicals
£22.30 - £63.20
Chemical Identifiers
CAS | 586-77-6 |
---|---|
Molecular Formula | C8H10BrN |
Molecular Weight (g/mol) | 200.08 |
MDL Number | MFCD00000093 |
InChI Key | XYZWMVYYUIMRIZ-UHFFFAOYSA-N |
Synonym | n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene |
PubChem CID | 11465 |
IUPAC Name | 4-bromo-N,N-dimethylaniline |
SMILES | CN(C)C1=CC=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499822
|
Thermo Scientific Alfa Aesar
A10854.14 |
25 g |
£22.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409832
|
Thermo Scientific Alfa Aesar
A10854.22 |
100 g |
£63.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and metals. Store at 4°C.
Chemical Identifiers
586-77-6 | |
200.08 | |
XYZWMVYYUIMRIZ-UHFFFAOYSA-N | |
11465 | |
CN(C)C1=CC=C(Br)C=C1 |
C8H10BrN | |
MFCD00000093 | |
n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene | |
4-bromo-N,N-dimethylaniline |
Specifications
586-77-6 | |
263°C to 265°C | |
C8H10BrN | |
2206155 | |
Soluble in methanol. Insoluble in water. | |
CN(C)C1=CC=C(Br)C=C1 | |
200.08 | |
200.08 | |
4-Bromo-N,N-dimethylaniline |
52°C to 56°C | |
>110°C (230°F) | |
MFCD00000093 | |
n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene | |
XYZWMVYYUIMRIZ-UHFFFAOYSA-N | |
4-bromo-N,N-dimethylaniline | |
11465 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 209-582-2
missing translation for 'rtecsNumber' : BW9300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only