missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenol, 98+%, Thermo Scientific Chemicals

Product Code. 11339429
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol

Specifications

Melting Point 118°C to 122°C
Density 1.26
Quantity 100 g
Beilstein 1958305
Merck Index 14,6248
Solubility Information Slightly soluble in water. Soluble in alcohol,acetone,benzene,and diethyl ether.
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O
Molecular Weight (g/mol) 200.237
Formula Weight 200.24
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'