Learn More
{4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine, 97%, Thermo Scientific™
£90.95 - £216.00
Chemical Identifiers
CAS | 906352-74-7 |
---|---|
Molecular Formula | C13H11F3N2 |
Molecular Weight (g/mol) | 252.24 |
MDL Number | MFCD09817478 |
InChI Key | XQAJYHFQYZEWSE-UHFFFAOYSA-N |
Synonym | 4-5-trifluoromethyl pyridin-2-yl benzylamine, 4-5-trifluoromethyl pyrid-2-yl phenyl methylamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methylamine, 1-4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl-2-pyridyl phenyl methylamine, 2-4-aminomethyl phenyl-5-trifluoromethyl pyridine |
PubChem CID | 24229534 |
IUPAC Name | [4-[5-(trifluoromethyl)pyridin-2-yl]phenyl]methanamine |
SMILES | C1=CC(=CC=C1CN)C2=NC=C(C=C2)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10608534
|
Thermo Scientific Maybridge
CC53013CB |
250 mg | Amber Glass Bottle |
£90.95
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10612675
|
Thermo Scientific Maybridge
CC53013DA |
1 g | Amber Glass Bottle |
£216.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
906352-74-7 | |
252.24 | |
XQAJYHFQYZEWSE-UHFFFAOYSA-N | |
24229534 | |
C1=CC(=CC=C1CN)C2=NC=C(C=C2)C(F)(F)F |
C13H11F3N2 | |
MFCD09817478 | |
4-5-trifluoromethyl pyridin-2-yl benzylamine, 4-5-trifluoromethyl pyrid-2-yl phenyl methylamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methylamine, 1-4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl-2-pyridyl phenyl methylamine, 2-4-aminomethyl phenyl-5-trifluoromethyl pyridine | |
[4-[5-(trifluoromethyl)pyridin-2-yl]phenyl]methanamine |
Specifications
906352-74-7 | |
100.0 | |
C13H11F3N2 | |
4-5-trifluoromethyl pyridin-2-yl benzylamine, 4-5-trifluoromethyl pyrid-2-yl phenyl methylamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methylamine, 1-4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl-2-pyridyl phenyl methylamine, 2-4-aminomethyl phenyl-5-trifluoromethyl pyridine | |
C1=CC(=CC=C1CN)C2=NC=C(C=C2)C(F)(F)F | |
252.24 | |
252.24 | |
{4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine |
96.5 | |
97% | |
MFCD09817478 | |
XQAJYHFQYZEWSE-UHFFFAOYSA-N | |
[4-[5-(trifluoromethyl)pyridin-2-yl]phenyl]methanamine | |
24229534 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger