missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Methyl-2-piperidinylsulfonyl)acetanilide, 97%, Thermo Scientific™

Product Code. 15485786
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858240-55-7
Molecular Formula C14H20N2O3S
Molecular Weight (g/mol) 296.39
MDL Number MFCD22683128
InChI Key LUPVXSOGPVCCJN-UHFFFAOYNA-N
Synonym n-4-4-methylpiperidin-2-ylsulfonyl phenyl acetamide, 4'-4-methyl-2-piperidinylsulfonyl acetanilide
PubChem CID 121237459
IUPAC Name N-[4-(4-methylpiperidin-2-yl)sulfonylphenyl]acetamide
SMILES CC1CCNC(C1)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1

Specifications

Quantity 1 g
Formula Weight 296.38
Percent Purity 97%
Chemical Name or Material 4'-(4-Methyl-2-piperidinylsulfonyl)acetanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'