missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chloro-4-methylphenylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15490296
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-37-7
Molecular Formula C14H13BClNO3
Molecular Weight (g/mol) 289.522
MDL Number MFCD09027211
InChI Key IZMAVYDCXUROFV-UHFFFAOYSA-N
Synonym n-3-chloro-4-methylphenyl 4-boronobenzamide, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronic acid, 4-3-chloro-4-methylphenyl carbamoyl benzeneboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenylboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronicacid, 4-3-chloro-4-methylphenyl carbamoyl-phenyl boronic acid, boronic acid,b-4-3-chloro-4-methylphenyl amino carbonyl phenyl
PubChem CID 44119159
IUPAC Name [4-[(3-chloro-4-methylphenyl)carbamoyl]phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC2=CC(=C(C=C2)C)Cl)(O)O

Specifications

Melting Point 217°C to 219°C
Quantity 1 g
Formula Weight 289.53
Percent Purity 98%
Chemical Name or Material 4-(3-Chloro-4-methylphenylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'