Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
Brand:  Greiner Bio-One 781904
 View more versions of this product
Product Code. 11893891
  • £258.40 / Pack of 40
Estimated Shipment: 22-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Not Autoclavable
White
Polystyrene
15 to 110 μL
131 μL
4800 x g (swinging bucket rotor)
10
-20°C to +60°C
16 x 24
Non Binding
Microplate
384
Non-sterile
Rounded Square
40
Yes
White
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate > White plate, F-bottom