missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3000 μS/cm Conductivity Standard at 25 °C, Reagecon
Aqueous based, non-hazardous standard
Brand: Reagecon CSKC3M
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Tested by ISO 17025 accredited test method
- NIST traceable
- Low temperature coefficient of variation
- Guaranteed stability throughout entire shelf life even after opening bottle
- Accuracy: 3000 μS/cm ± 1%
Specifications
HDPE bottle GL45 | |
Yes | |
Conductivity Standard | |
Liquid |
500 mL | |
Yes | |
Conductivity Meter |