Learn More
3-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
£13.90 - £198.00
Chemical Identifiers
CAS | 121-90-4 |
---|---|
Molecular Formula | C7H4ClNO3 |
Molecular Weight (g/mol) | 185.57 |
MDL Number | MFCD00007247 |
InChI Key | NXTNASSYJUXJDV-UHFFFAOYSA-N |
Synonym | m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid |
PubChem CID | 8495 |
IUPAC Name | 3-nitrobenzoyl chloride |
SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10327810
|
Thermo Scientific Acros
155840050 |
5 g | Glass bottle | ||||||
10512751
|
Thermo Scientific Acros
155841000 |
100 g | Glass bottle | ||||||
10153490
|
Thermo Scientific Acros
155845000 |
500 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
121-90-4 | |
185.57 | |
NXTNASSYJUXJDV-UHFFFAOYSA-N | |
8495 | |
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl |
C7H4ClNO3 | |
MFCD00007247 | |
m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid | |
3-nitrobenzoyl chloride |
Specifications
121-90-4 | |
100.0 | |
Brown | |
275.0°C to 278.0°C | |
Authentic | |
C7H4ClNO3 | |
MFCD00007247 | |
1.42 | |
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol | |
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl | |
185.57 | |
185.57 | |
Low Melting Solid |
97.5 | |
30.0°C to 35.0°C | |
1.4200g/mL | |
>112°C | |
98% | |
O2NC6H4COCl | |
09, 381 | |
m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid | |
NXTNASSYJUXJDV-UHFFFAOYSA-N | |
3-nitrobenzoyl chloride | |
8495 | |
98% | |
3-Nitrobenzoyl chloride, 98% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-505-9
missing translation for 'rtecsNumber' : DM6646000
missing translation for 'tsca' : TSCA
RUO – Research Use Only