Learn More
3-Methylbenzyl bromide, 97%, Thermo Scientific Chemicals
£23.72 - £65.50
Chemical Identifiers
CAS | 620-13-3 |
---|---|
Molecular Formula | C8H9Br |
Molecular Weight (g/mol) | 185.064 |
MDL Number | MFCD00000177 |
InChI Key | FWLWTILKTABGKQ-UHFFFAOYSA-N |
Synonym | 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo |
PubChem CID | 12099 |
IUPAC Name | 1-(bromomethyl)-3-methylbenzene |
SMILES | CC1=CC(=CC=C1)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11426426
|
Thermo Scientific Alfa Aesar
B24731.06 |
5 g |
£23.72
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436426
|
Thermo Scientific Alfa Aesar
B24731.14 |
25 g |
£65.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
620-13-3 | |
185.064 | |
FWLWTILKTABGKQ-UHFFFAOYSA-N | |
12099 | |
CC1=CC(=CC=C1)CBr |
C8H9Br | |
MFCD00000177 | |
3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
1-(bromomethyl)-3-methylbenzene |
Specifications
620-13-3 | |
184°C to 185°C (340 mmHg) | |
C8H9Br | |
MFCD00000177 | |
2076425 | |
3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
CC1=CC(=CC=C1)CBr | |
185.064 | |
185.07 | |
3-Methylbenzyl bromide |
1.37 | |
82°C (179°F) | |
1.569 | |
UN1701 | |
14,10090 | |
FWLWTILKTABGKQ-UHFFFAOYSA-N | |
1-(bromomethyl)-3-methylbenzene | |
12099 | |
97% |
Safety and Handling
GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLYL BROMIDE, LIQUID
missing translation for 'einecsNumber' : 210-625-2
missing translation for 'rtecsNumber' : CY9039200
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only