missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-cyclopentanedione, 99%, Thermo Scientific Chemicals

Product Code. 10720955
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-70-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001417
InChI Key OACYKCIZDVVNJL-UHFFFAOYSA-N
Synonym 3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
PubChem CID 61209
IUPAC Name 3-methylcyclopentane-1,2-dione
SMILES CC1CCC(=O)C1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 105.0°C to 110.0°C
Color White
Density 100g/mL
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 250 g
Beilstein 07, I, 310
Solubility Information Solubility in water: soluble.
Formula Weight 112.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Methyl-1,2-cyclopentanedione
Show More Show Less
Product Identifier
  • 3-Methyl-1,2-cyclopentanedione

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'