missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
3-Methoxybenzylmagnesium chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxybenzylmagnesium chloride is used as pharmaceutical intermediate.
Solubility
It reacts with water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
3-Methoxybenzylmagnesium chloride is used as pharmaceutical intermediate.
Solubility
It reacts with water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications
CAS | 26905-40-8 |
Color | Brown to Gray |
Molecular Formula | C8H9ClMgO |
MDL Number | MFCD01319896 |
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Synonym | 3-methoxybenzylmagnesium chloride, 3-methoxybenzyl magnesiumchloride, 3-methoxybenzyl magnesium chloride, zpcuemlxjgelgi-uhfffaoysa-m, magnesium,chloro 3-methoxyphenyl methyl, 3-methoxybenzylmagnesium chloride 0.25 m in tetrahydrofuran, 3-methoxybenzylmagnesium chloride solution, 0.25 m in thf |
Solubility Information | It reacts with water. |
InChI Key | ZPCUEMLXJGELGI-UHFFFAOYSA-M |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'