missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-2-methylpropyl chloride hydrochloride, 98%, Thermo Scientific™

Product Code. 11352366
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4261-67-0
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.093
MDL Number MFCD00012514
InChI Key SOMIBONUMGNAEP-UHFFFAOYSA-N
Synonym 3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
PubChem CID 3083841
IUPAC Name 3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)CCl.Cl

Specifications

Melting Point 167°C to 170°C
Quantity 100 g
Beilstein 4444785
Sensitivity Hygroscopic
Formula Weight 172.1
Percent Purity 98%
Chemical Name or Material 3-Dimethylamino-2-methylpropyl chloride hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'