missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzeneboronic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11395425
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150255-96-2
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318967
InChI Key XDBHWPLGGBLUHH-UHFFFAOYSA-N
Synonym 3-cyanophenyl boronic acid, 3-cyanobenzeneboronic acid, 3-boronobenzonitrile, 3-dihydroxyboranyl benzonitrile, 3-cyanobenzene boronic acid, 3-cyano-phenyl-boronic acid, boronic acid, 3-cyanophenyl, pubchem1804, phenylboronic acid, 4
PubChem CID 2734325
IUPAC Name (3-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)C#N

Specifications

Melting Point ∽298°C (decomposition)
Quantity 250 mg
Beilstein 6136720
Formula Weight 146.94
Percent Purity ≥98%
Chemical Name or Material 3-Cyanobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'