Learn More
3-Chloro-6-iodopyridazine, 95%, Thermo Scientific Chemicals
£49.45 - £179.00
Chemical Identifiers
CAS | 135034-10-5 |
---|---|
Molecular Formula | C4H2ClIN2 |
Molecular Weight (g/mol) | 240.43 |
MDL Number | MFCD08275187 |
InChI Key | PNEPCDPKMXJYIQ-UHFFFAOYSA-N |
Synonym | 3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine |
PubChem CID | 15418839 |
IUPAC Name | 3-chloro-6-iodopyridazine |
SMILES | ClC1=NN=C(I)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15400829
|
Thermo Scientific Alfa Aesar
H63879.03 |
1 g |
£49.45
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15410829
|
Thermo Scientific Alfa Aesar
H63879.06 |
5 g |
£179.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
Chemical Identifiers
135034-10-5 | |
240.43 | |
PNEPCDPKMXJYIQ-UHFFFAOYSA-N | |
15418839 | |
ClC1=NN=C(I)C=C1 |
C4H2ClIN2 | |
MFCD08275187 | |
3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine | |
3-chloro-6-iodopyridazine |
Specifications
135034-10-5 | |
C4H2ClIN2 | |
Light sensitive | |
Slightly soluble in water. | |
ClC1=NN=C(I)C=C1 | |
240.43 | |
240.43 | |
Solid |
110°C to 120°C | |
MFCD08275187 | |
3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine | |
PNEPCDPKMXJYIQ-UHFFFAOYSA-N | |
3-chloro-6-iodopyridazine | |
15418839 | |
97% | |
3-Chloro-6-iodopyridazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only