missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxy-5-methoxybenzyl alcohol, 98%, Thermo Scientific™

Product Code. 11416147
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20624-92-4
Molecular Formula C8H9ClO3
Molecular Weight (g/mol) 188.61
MDL Number MFCD00016870
InChI Key VQEZBFOBPUKDGM-UHFFFAOYSA-N
Synonym 5-chlorovanillyl alcohol, 2-chloro-4-hydroxymethyl-6-methoxyphenol, 3-chloro-4-hydroxy-5-methoxybenzyl alcohol, 3-chloro-4-hydroxy-5-methoxybenzenemethanol, acmc-20am3y, 6-chloro-4-hydroxymethyl-2-methoxyphenol, 5-chloro-4-hydroxy-3-methoxybenzenemethanol, benzenemethanol,3-chloro-4-hydroxy-5-methoxy
PubChem CID 4181655
IUPAC Name 2-chloro-4-(hydroxymethyl)-6-methoxyphenol
SMILES COC1=C(O)C(Cl)=CC(CO)=C1

Specifications

Melting Point 115°C to 118°C
Quantity 25 g
Formula Weight 188.62
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxybenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'