missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-benzisothiazole, 97+%, Thermo Scientific Chemicals

Product Code. 11304765
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7716-66-7
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00673254
InChI Key BCPVKLRBQLRWDQ-UHFFFAOYSA-N
Synonym 3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole
PubChem CID 598190
IUPAC Name 3-chloro-1,2-benzothiazole
SMILES C1=CC=C2C(=C1)C(=NS2)Cl

Specifications

Melting Point 37°C to 44°C
Boiling Point 80°C to 86°C (0.75 mmHg)
Quantity 25 g
Beilstein 119370
Solubility Information Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
Formula Weight 169.63
Percent Purity 99%
Chemical Name or Material 3-Chloro-1,2-benzisothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'