missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Bromomethyl)-5-chlorobenzo[b]thiophene, 95+%, Thermo Scientific™
£10.40 - £52.95
Chemical Identifiers
CAS | 1198-51-2 |
---|---|
Molecular Formula | C9H6BrClS |
Molecular Weight (g/mol) | 261.56 |
MDL Number | MFCD00173769 |
InChI Key | FKQSFVITUNJLCY-UHFFFAOYSA-N |
Synonym | 3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553 |
PubChem CID | 2779857 |
IUPAC Name | 3-(bromomethyl)-5-chloro-1-benzothiophene |
SMILES | ClC1=CC=C2SC=C(CBr)C2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10713633
|
Thermo Scientific Maybridge
MWP00055DA |
1 g | Amber Glass Bottle |
£10.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386944
|
Thermo Scientific Maybridge
MWP00055DE |
5 g | Amber Glass Bottle |
£28.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205844
|
Thermo Scientific Maybridge
MWP00055EA |
10 g | Amber Glass Bottle |
£52.95
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1198-51-2 | |
261.56 | |
FKQSFVITUNJLCY-UHFFFAOYSA-N | |
2779857 | |
ClC1=CC=C2SC=C(CBr)C2=C1 |
C9H6BrClS | |
MFCD00173769 | |
3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553 | |
3-(bromomethyl)-5-chloro-1-benzothiophene |
Specifications
1198-51-2 | |
C9H6BrClS | |
3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553 | |
ClC1=CC=C2SC=C(CBr)C2=C1 | |
261.56 | |
261.57 | |
3-(Bromomethyl)-5-chlorobenzo[b]thiophene |
95% | |
MFCD00173769 | |
FKQSFVITUNJLCY-UHFFFAOYSA-N | |
3-(bromomethyl)-5-chloro-1-benzothiophene | |
2779857 | |
95% |