missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals

Product Code. 11489973
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

Solubility
Soluble in water.

Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-18-8
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.10
MDL Number MFCD00014820
InChI Key AGSPXMVUFBBBMO-UHFFFAOYSA-N
Synonym 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino
PubChem CID 1647
ChEBI CHEBI:27413
IUPAC Name 3-aminopropanenitrile
SMILES NCCC#N

Specifications

Density 0.952
Boiling Point 79°C to 81°C (16 mmHg)
Odor Amine-like
Linear Formula H2NCH2CH2CN
Refractive Index 1.4375
Quantity 5 g
UN Number UN2735
Beilstein 1698848
Sensitivity Air and moisture sensitive
Merck Index 14,469
Solubility Information Soluble in water.
Formula Weight 70.1
Percent Purity 98%
Chemical Name or Material 3-Aminopropionitrile, stabilized with potassium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'