missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11478826
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.

Solubility
Soluble in water, dimethyl sulfoxide, methanol, ethanol, diethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-99-4
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.116
MDL Number MFCD00007104
InChI Key LWFUFLREGJMOIZ-UHFFFAOYSA-N
Synonym 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid
PubChem CID 11873
ChEBI CHEBI:53648
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 169°C to 174°C
Quantity 1000 g
Beilstein 2220661
Solubility Information Soluble in water,dimethyl sulfoxide,methanol,ethanol,diethyl ether and benzene.
Formula Weight 228.12
Percent Purity ≥97%
Chemical Name or Material 3,5-Dinitrosalicylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'