missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxybenzoyl chloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents. 3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift. It is used as a chemical and organic intermediate.
Solubility
It hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents. 3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift. It is used as a chemical and organic intermediate.
Solubility
It hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
Specifications
Specifications
Melting Point | 42°C to 44°C |
Boiling Point | 152°C to 154°C (11 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 511839 |
Sensitivity | Moisture sensitive |
Synonym | benzoyl chloride, 3,5-dimethoxy, 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-benzoyl chloride, acmc-1c560, benzoyl chloride,3,5-dimethoxy, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride |
Solubility Information | It hydrolyzes in water. |
Formula Weight | 200.62 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'