missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzamide, 97%, Thermo Scientific™

Product Code. 11398175
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3147-62-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00017123
InChI Key PLYYMFBDRBSPJZ-UHFFFAOYSA-N
Synonym benzamide, 3,5-dihydroxy, 3,5-dihydroxy-benzamide, alpha-resorcylamide, 3,5-resorcylic acidamide, 3,5-dihydroxybenzoic acid amide, .alpha.-resorcylamide, pubchem13928, a-resorcyl amide, acmc-1coua, ksc494k8l
PubChem CID 76604
IUPAC Name 3,5-dihydroxybenzamide
SMILES C1=C(C=C(C=C1O)O)C(=O)N

Specifications

Melting Point ∽280°C (decomposition)
Quantity 5 g
Beilstein 2690560
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 3,5-Dihydroxybenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'