missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluoro-4-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
£107.00 - £219.00
Chemical Identifiers
CAS | 1029439-83-5 |
---|---|
Molecular Formula | C12H15BF2O3 |
Molecular Weight (g/mol) | 256.056 |
MDL Number | MFCD16996286 |
InChI Key | KTQYJNVSVKKLGQ-UHFFFAOYSA-N |
Synonym | 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3,5-difluoro-4-hydroxybenzeneboronic acid pinacol ester, 3,5-difluoro-4-hydroxyphenylboronic acid pinacol ester |
PubChem CID | 57655406 |
IUPAC Name | 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)O)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426966
|
Thermo Scientific Alfa Aesar
H62042.MD |
250 mg |
£107.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416966
|
Thermo Scientific Alfa Aesar
H62042.03 |
1 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1029439-83-5 | |
256.056 | |
KTQYJNVSVKKLGQ-UHFFFAOYSA-N | |
57655406 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)O)F |
C12H15BF2O3 | |
MFCD16996286 | |
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3,5-difluoro-4-hydroxybenzeneboronic acid pinacol ester, 3,5-difluoro-4-hydroxyphenylboronic acid pinacol ester | |
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
Specifications
1029439-83-5 | |
C12H15BF2O3 | |
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3,5-difluoro-4-hydroxybenzeneboronic acid pinacol ester, 3,5-difluoro-4-hydroxyphenylboronic acid pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)O)F | |
256.056 | |
256.05 | |
Solid |
White | |
MFCD16996286 | |
KTQYJNVSVKKLGQ-UHFFFAOYSA-N | |
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
57655406 | |
96% | |
3,5-Difluoro-4-hydroxybenzeneboronic acid pinacol ester |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only