Learn More
3,4,5-Trimethoxyphenol, 97%, Thermo Scientific Chemicals
£41.70 - £128.00
Chemical Identifiers
CAS | 642-71-7 |
---|---|
Molecular Formula | C9H12O4 |
Molecular Weight (g/mol) | 184.191 |
MDL Number | MFCD00008389 |
InChI Key | VTCDZPUMZAZMSB-UHFFFAOYSA-N |
Synonym | antiarol, phenol, 3,4,5-trimethoxy, 3,4,5-trimethoxy phenol, spectrum_000525, specplus_000623, spectrum3_001208, spectrum4_001487, spectrum5_000193, 3,4,5-trirnethoxyphenol, 3,4,5-trimethoxy-phenol |
PubChem CID | 69505 |
ChEBI | CHEBI:2760 |
IUPAC Name | 3,4,5-trimethoxyphenol |
SMILES | COC1=CC(=CC(=C1OC)OC)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311317
|
Thermo Scientific Alfa Aesar
L01527.06 |
5 g |
£41.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321317
|
Thermo Scientific Alfa Aesar
L01527.14 |
25 g |
£128.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.
Solubility
Soluble in chloroform.
Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
Chemical Identifiers
642-71-7 | |
184.191 | |
VTCDZPUMZAZMSB-UHFFFAOYSA-N | |
69505 | |
3,4,5-trimethoxyphenol |
C9H12O4 | |
MFCD00008389 | |
antiarol, phenol, 3,4,5-trimethoxy, 3,4,5-trimethoxy phenol, spectrum_000525, specplus_000623, spectrum3_001208, spectrum4_001487, spectrum5_000193, 3,4,5-trirnethoxyphenol, 3,4,5-trimethoxy-phenol | |
CHEBI:2760 | |
COC1=CC(=CC(=C1OC)OC)O |
Specifications
642-71-7 | |
C9H12O4 | |
1958573 | |
antiarol, phenol, 3,4,5-trimethoxy, 3,4,5-trimethoxy phenol, spectrum_000525, specplus_000623, spectrum3_001208, spectrum4_001487, spectrum5_000193, 3,4,5-trirnethoxyphenol, 3,4,5-trimethoxy-phenol | |
VTCDZPUMZAZMSB-UHFFFAOYSA-N | |
3,4,5-trimethoxyphenol | |
69505 | |
184.19 | |
3,4,5-Trimethoxyphenol |
143°C to 148°C | |
MFCD00008389 | |
Light sensitive | |
Soluble in chloroform. | |
COC1=CC(=CC(=C1OC)OC)O | |
184.191 | |
CHEBI:2760 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 211-387-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only