missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trihydroxybenzoic acid monohydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It's also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It′s also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.
Solubility
It is soluble in alcohol, ether, glycerol, acetone negligible in benzene, chloroform, petroleum ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Light sensitive. Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides.
This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It′s also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.
Solubility
It is soluble in alcohol, ether, glycerol, acetone negligible in benzene, chloroform, petroleum ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Light sensitive. Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides.
Specifications
Melting Point | 250°C (decomposition) |
Quantity | 250 g |
Sensitivity | Light sensitive |
Merck Index | 144345 |
Solubility Information | It is soluble in alcohol,ether,glycerol,acetone negligible in benzene,chloroform,petroleum ether. |
Formula Weight | 188.14 (170.12 Anhydrous) |
Percent Purity | 0.99 |
Chemical Name or Material | 3,4,5-Trihydroxybenzoic acid monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'