missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7027844
Click to view available options
Quantity:
250 g
1000 g
149-91-7
C7H6O5
170.12
MFCD00002510
LNTHITQWFMADLM-UHFFFAOYSA-N
gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
370
CHEBI:30778
3,4,5-trihydroxybenzoic acid
OC(=O)C1=CC(O)=C(O)C(O)=C1
149-91-7
C7H6O5
170.12
MFCD00002510
LNTHITQWFMADLM-UHFFFAOYSA-N
gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
370
CHEBI:30778
3,4,5-trihydroxybenzoic acid
OC(=O)C1=CC(O)=C(O)C(O)=C1

Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

Solubility
Soluble in water, alcohol, ether, glycerol and acetone. Insoluble in benzene, chloroform and petroleum ether.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents, strong bases, acid anhydrides, acid chlorides, metals.
TRUSTED_SUSTAINABILITY
Melting Point ∼251°C (decomposition)
Density 1.694
Quantity 250 g
Sensitivity Hygroscopic; Light sensitive
Merck Index 144345
Solubility Information Soluble in water,alcohol,ether,glycerol and acetone. Insoluble in benzene,chloroform and petroleum ether.
Formula Weight 170.12
Percent Purity 0.99
Chemical Name or Material 3,4,5-Trihydroxybenzoic acid, anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'