missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Stopcocks

£24.00 - £248.00

Specifications

Type Stopcock
Material Borosilicate Glass
View More Specs

Products 6
Product Code Brand Includes Plug Bore Description Price Quantity & Availability  
Product Code Brand Includes Plug Bore Description Price Quantity & Availability  
10499162
missing translation for 'viewDocuments' Promotion Details
Lenz Laborglasinstrumente™
02161202
PTFE plug and screw thread retaining device 2.5 mm Plug size: NS 12.5
£227.00
Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
10622571
missing translation for 'viewDocuments' Promotion Details
Lenz Laborglasinstrumente™
02221402
Solid Glass plug, capillary side arms 2.5 mm Plug size: NS 14.5
£248.00
Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
10160581
missing translation for 'viewDocuments' Promotion Details
Lenz Laborglasinstrumente™
02101202
Solid Glass plug 2.5 mm Plug size: NS 12.5
£129.00
Pack of 10
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
10778711
missing translation for 'viewDocuments' Promotion Details
Lenz Laborglasinstrumente™
02101404
Solid Glass plug 4 mm Plug size: NS 14.5
£191.00
Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
10728141
missing translation for 'viewDocuments' Promotion Details
Lenz Laborglasinstrumente™
02161804
PTFE plug and screw thread retaining device 4 mm Plug size: NS 18.8
£35.00
Each
Estimated Shipment: 21-11-2024
to see stock.
Add to basket
 
10489742
missing translation for 'viewDocuments' Promotion Details
Lenz Laborglasinstrumente™
02101804
Solid Glass plug 4 mm Plug size: NS 18.8
£24.00
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Stopcock
Borosilicate Glass
Videos
SDS
Documents

Documents

Certificates