missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Pyrrolyl)-2-thiophenemethanol, 97%, Thermo Scientific™

Product Code. 15487177
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107073-27-8
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00052578
InChI Key SMSUHWYHSLZKHS-UHFFFAOYSA-N
Synonym 3-pyrrol-1-yl thiophene-2-methanol, 3-1h-pyrrol-1-yl-2-thienyl methanol, 3-pyrrol-1-ylthiophen-2-yl methanol, 3-1h-pyrrol-1-yl thiophen-2-yl methanol, 3-pyrrol-1-yl thiophen-2-yl methanol, 3-1h-pyrrol-1-yl thien-2-yl methanol, 3-pyrrolyl-2-thienyl methan-1-ol, 3-1-pyrrolyl-2-thiophenemethanol, 3-1-pyrrolyl-2-thiophenyl methanol
PubChem CID 342464
IUPAC Name (3-pyrrol-1-ylthiophen-2-yl)methanol
SMILES C1=CN(C=C1)C2=C(SC=C2)CO

Specifications

Boiling Point 104°C to 108°C (1.5 mmHg)
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 3-(1-Pyrrolyl)-2-thiophenemethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'