missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Azepanyl)-2,2-dimethylpropylamine, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 184.32 |
Percent Purity | 95% |
Chemical Name or Material | 3-(1-Azepanyl)-2, 2-dimethylpropylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'