Learn More
3M™ Polycarbonate Protective Safety Glasses, 2740 Series
Click to view available options
Lens Tint:
Amber
Clear
Gray
Description
Additional features and style increase security, comfort, and wearer acceptance.
- Adjustable temple length to suit the individual
- Excellent impact resistance
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Anti-scratch and anti-fog coating for increased durability and improved vision
- Optical Class 1 for prolonged use
- Ideal for use in many environments including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Amber |
Product Type | Safety glasses; Safety Spectacles |
For Use With (Application) | Construction, Metal fabrication, Forestry |
Frame Color | Blue |
No. per Pack | 1 Ea. |
Temple Style | Adjustable length, comfortable fit |
Lens Material | Polycarbonate |
Product Content Correction
Product Title
3M™ Polycarbonate Protective Safety Glasses, 2740 Series >
Spot an opportunity for improvement?Share a Content Correction