missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11369725
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175676-65-0
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320763
InChI Key AIJCNTOYZPKURP-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
PubChem CID 2777204
IUPAC Name [2-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1OC(F)(F)F

Specifications

Melting Point 118°C to 120°C
Quantity 1 g
Beilstein 8766167
Formula Weight 205.93
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethoxy)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'