missing translation for 'onlineSavingsMsg'
Learn More

2-Oxocyclohexanepropionitrile, 99%, Thermo Scientific Chemicals

Product Code. 11404640
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4594-78-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00013797
InChI Key SPTVCXKSSRBTMN-UHFFFAOYNA-N
Synonym 3-2-oxocyclohexyl propanenitrile, 2-2-cyanoethyl cyclohexanone, cyclohexanepropanenitrile, 2-oxo, 2-2-cyanethyl cyclohexanone, 2-oxo-1-cyclohexanepropionitrile, cyclohexanepropionitrile, 2-oxo, 2-oxocyclohexanepropionitrile, 2-.beta.-cyanoethyl cyclohexanone, 2-b-cyanoethyl cyclohexanone, 2-oxocyclohexanepropiononitrile
PubChem CID 96223
IUPAC Name 3-(2-oxocyclohexyl)propanenitrile
SMILES O=C1CCCCC1CCC#N

Specifications

Density 1.021
Boiling Point 138°C to 142°C (10 mmHg)
Quantity 5 g
UN Number UN3276
Formula Weight 151.21
Percent Purity 99%
Chemical Name or Material 2-Oxocyclohexanepropionitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'