missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine, 96%, Thermo Scientific™

Catalog Number p-7034014
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
122476-85-1
C17H32N2O2SSn
447.225
MFCD07787379
RRHTVQOGUAOHDR-UHFFFAOYSA-N
2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine
2763272
tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C
This item is not returnable. View return policy
122476-85-1
C17H32N2O2SSn
447.225
MFCD07787379
RRHTVQOGUAOHDR-UHFFFAOYSA-N
2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine
2763272
tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2788
Formula Weight 447.22
Percent Purity 96%
Chemical Name or Material 2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'