Learn More
2-Methylcyclopentane-1,3-dione, 98%, Thermo Scientific Chemicals
£27.40 - £330.00
Chemical Identifiers
CAS | 765-69-5 |
---|---|
Molecular Formula | C6H8O2 |
Molecular Weight (g/mol) | 112.128 |
MDL Number | MFCD00001406 |
InChI Key | HXZILEQYFQYQCE-UHFFFAOYSA-N |
Synonym | 2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46 |
PubChem CID | 13005 |
IUPAC Name | 2-methylcyclopentane-1,3-dione |
SMILES | CC1C(=O)CCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11411668
|
Thermo Scientific Alfa Aesar
A13936.06 |
5 g |
£27.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11421668
|
Thermo Scientific Alfa Aesar
A13936.14 |
25 g |
£88.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431668
|
Thermo Scientific Alfa Aesar
A13936.22 |
100 g |
£330.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.
Solubility
Solubility in hot Methanol (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
765-69-5 | |
112.128 | |
HXZILEQYFQYQCE-UHFFFAOYSA-N | |
13005 | |
CC1C(=O)CCC1=O |
C6H8O2 | |
MFCD00001406 | |
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46 | |
2-methylcyclopentane-1,3-dione |
Specifications
765-69-5 | |
C6H8O2 | |
1237255 | |
HXZILEQYFQYQCE-UHFFFAOYSA-N | |
2-methylcyclopentane-1,3-dione | |
13005 | |
98% |
212°C to 216°C | |
MFCD00001406 | |
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46 | |
CC1C(=O)CCC1=O | |
112.128 | |
112.13 | |
2-Methylcyclopentane-1,3-dione |
Safety and Handling
H500
missing translation for 'einecsNumber' : 212-153-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only