Learn More
2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals
£103.00 - £1176.00
Chemical Identifiers
CAS | 583-60-8 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.172 |
MDL Number | MFCD00001635 |
InChI Key | LFSAPCRASZRSKS-UHFFFAOYSA-N |
Synonym | 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone |
PubChem CID | 11419 |
IUPAC Name | 2-methylcyclohexan-1-one |
SMILES | CC1CCCCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359226
|
Thermo Scientific Alfa Aesar
A14741.AE |
100 mL |
£103.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403097
|
Thermo Scientific Alfa Aesar
A14741.AP |
500 mL |
£338.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369226
|
Thermo Scientific Alfa Aesar
A14741.0F |
2500 mL |
£1176.00
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.
Solubility
Insoluble in water. Soluble in methanol and diethyl ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
583-60-8 | |
112.172 | |
LFSAPCRASZRSKS-UHFFFAOYSA-N | |
11419 | |
CC1CCCCC1=O |
C7H12O | |
MFCD00001635 | |
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone | |
2-methylcyclohexan-1-one |
Specifications
583-60-8 | |
0.925 | |
46°C (114°F) | |
C7H12O | |
MFCD00001635 | |
506751 | |
Insoluble in water. Soluble in methanol and diethyl ether. | |
CC1CCCCC1=O | |
112.172 | |
112.17 | |
Liquid |
°C to 14°C | |
162°C to 163°C | |
98% | |
1.448 | |
UN2297 | |
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone | |
LFSAPCRASZRSKS-UHFFFAOYSA-N | |
2-methylcyclohexan-1-one | |
11419 | |
98% | |
2-Methylcyclohexanone |
Safety and Handling
GHS H Statement
H226-H332
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYLCYCLOHEXANONE
missing translation for 'einecsNumber' : 209-513-6
missing translation for 'rtecsNumber' : GW1750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only