missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-Propanol, Honeywell
£82.64 - £163.37
Chemical Identifiers
CAS | 78-83-1 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00004740 |
InChI Key | ZXEKIIBDNHEJCQ-UHFFFAOYSA-N |
Synonym | 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol |
PubChem CID | 6560 |
ChEBI | CHEBI:46645 |
IUPAC Name | 2-methylpropan-1-ol |
SMILES | CC(C)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633880
|
Honeywell Riedel-de Haën
33064-1L |
1 L |
£82.64
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643880
|
Honeywell Riedel-de Haën
33064-2.5L |
2.5 L |
£163.37
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
78-83-1 | |
74.123 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
6560 | |
2-methylpropan-1-ol |
C4H10O | |
MFCD00004740 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CHEBI:46645 | |
CC(C)CO |
Specifications
78-83-1 | |
28°C (82.4°F) | |
(CH3)2CHCH2OH | |
1212 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CC(C)CO | |
74.123 | |
2.55 (vs air) | |
74.12g/mol | |
ACS/puriss. p.a. |
0.803g/mL at 25°C | |
C4H10O | |
MFCD00004740 | |
1730878 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
2-methylpropan-1-ol | |
6560 | |
CHEBI:46645 | |
≥99% (GC) | |
2-Methyl-1-Propanol, Reag. Ph. Eur. |
Safety and Handling
P210-P280-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
H226-H315-H318-H335-H336
Danger
missing translation for 'einecsNumber' : 201-148-0
missing translation for 'rtecsNumber' : NP9625000