missing translation for 'onlineSavingsMsg'
Learn More

2-Mesitylethanol, 98%, Thermo Scientific™

Product Code. 11397955
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6950-92-1
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00020609
InChI Key FQZPTDPHVFTOSY-UHFFFAOYSA-N
Synonym 2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol
PubChem CID 81389
IUPAC Name 2-(2,4,6-trimethylphenyl)ethanol
SMILES CC1=CC(C)=C(CCO)C(C)=C1

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Beilstein 1938400
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 2-Mesitylethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'