missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11402408
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-13-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003332
InChI Key FZHSPPYCNDYIKD-UHFFFAOYSA-N
Synonym 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
PubChem CID 95695
IUPAC Name 2-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)O)C=O

Specifications

Melting Point 13°C to 16°C
Density 1.221
Boiling Point 248°C to 252°C
Flash Point >110°C (230°F)
Refractive Index 1.578
Quantity 100 g
Beilstein 1100532
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform. Slightly miscible with water.
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'