missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-2698443
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
1993-03-9
C6H6BFO2
139.92
MFCD00674013
QCSLIRFWJPOENV-UHFFFAOYSA-N
2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl, boronic acid, 2-fluorophenyl, 2-fluoro-phenyl-boronic acid, o-fluoro-benzeneboronic acid, benzeneboronic acid, o-fluoro
2734354
(2-fluorophenyl)boronic acid
OB(O)C1=CC=CC=C1F
This item is not returnable. View return policy
1993-03-9
C6H6BFO2
139.92
MFCD00674013
QCSLIRFWJPOENV-UHFFFAOYSA-N
2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl, boronic acid, 2-fluorophenyl, 2-fluoro-phenyl-boronic acid, o-fluoro-benzeneboronic acid, benzeneboronic acid, o-fluoro
2734354
(2-fluorophenyl)boronic acid
OB(O)C1=CC=CC=C1F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 139.92
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'