missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15484886
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1029439-02-8
Molecular Formula C12H16BFO3
Molecular Weight (g/mol) 238.065
MDL Number MFCD16994421
InChI Key ASYQJYIEVNXWDV-UHFFFAOYSA-N
Synonym 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-fluoro-4-hydroxybenzeneboronic acid pinacol ester, 2-fluoro-4-hydroxyphenylboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenol, phenol, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol
PubChem CID 54759098
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)O)F

Specifications

Melting Point 110°C to 112°C
Quantity 1 g
Formula Weight 238.07
Percent Purity 96%
Chemical Name or Material 2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'