Learn More
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
£40.40 - £405.00
Chemical Identifiers
CAS | 31106-82-8 |
---|---|
Molecular Formula | C6H6BrN·HBr |
Molecular Weight (g/mol) | 252.94 |
MDL Number | MFCD01863544 |
InChI Key | JQDNCGRNPYKRAO-UHFFFAOYSA-N |
Synonym | 2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide |
PubChem CID | 2734720 |
IUPAC Name | 2-(bromomethyl)pyridine;hydrobromide |
SMILES | C1=CC=NC(=C1)CBr.Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10581892
|
Thermo Scientific Acros
370110050 |
5 g | Glass bottle |
£40.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10794753
|
Thermo Scientific Acros
370110250 |
25 g | Glass bottle |
£132.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606884
|
Thermo Scientific Acros
370111000 |
100 g | Glass bottle |
£405.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31106-82-8 | |
252.94 | |
JQDNCGRNPYKRAO-UHFFFAOYSA-N | |
2734720 | |
C1=CC=NC(=C1)CBr.Br |
C6H6BrN·HBr | |
MFCD01863544 | |
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide | |
2-(bromomethyl)pyridine;hydrobromide |
Specifications
31106-82-8 | |
100.0 | |
Brown to White | |
98% | |
BrCH2(C5H3N) | |
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide | |
C1=CC=NC(=C1)CBr.Br | |
252.94 | |
252.94 | |
Crystalline Powder or Crystals |
97.5 | |
149.0°C to 152.0°C | |
Authentic | |
C6H6BrN·HBr | |
MFCD01863544 | |
JQDNCGRNPYKRAO-UHFFFAOYSA-N | |
2-(bromomethyl)pyridine;hydrobromide | |
2734720 | |
98% | |
2-(Bromomethyl)pyridine hydrobromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
- 2-(Bromomethyl)pyridine hydrobromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
RUO – Research Use Only