missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyrimidine, 97%, Thermo Scientific Chemicals

Product Code. 15440788
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130645-48-6
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD04037883
InChI Key UZIJEBOLOXOVFY-UHFFFAOYSA-N
PubChem CID 10942928
IUPAC Name 2-bromo-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Br

Specifications

Melting Point 45°C to 50°C
Quantity 5 g
Synonym 2-bromo-4-methyl-pyrimidine, pyrimidine, 2-bromo-4-methyl, pyrimidine,2-bromo-4-methyl, pubchem7007, acmc-209z1w, 2-bromo-4-methlypyrimidine, 2-bromo-4-methyl pyrimidine, 2-bromo-4-methylpyrimidine
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylpyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'