missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylaminopyridine, 98%, Thermo Scientific™

Product Code. 11450587
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6935-27-9
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00006251
InChI Key WYHXNQXDQQMTQI-UHFFFAOYSA-N
Synonym 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g
PubChem CID 23362
IUPAC Name N-benzylpyridin-2-amine
SMILES C1=CC=C(C=C1)CNC2=CC=CC=N2

Specifications

Melting Point 93°C to 97°C
Boiling Point 182°C (12 mmHg)
Flash Point 153°C (307°F)
Quantity 1000 g
Beilstein 134309
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material 2-Benzylaminopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'