missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminothiazole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2500 g
Description
2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
Soluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 88°C to 92°C |
Boiling Point | 117°C (15 mmHg) |
Quantity | 50 g |
Beilstein | 105738 |
Merck Index | 14,479 |
Solubility Information | Soluble in water,alcohol and ether. |
Formula Weight | 100.14 |
Percent Purity | 97% |
Chemical Name or Material | 2-Aminothiazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'